siRNA Details
| Gene Symbol: Kmt2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033276.3 | NP_001028448.3 | 51 | 16321 | ||
| GenBank mRNA | BC058659.1 | - | 12 | 3080 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kmt2d| Gene Symbol: Kmt2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033276.3 | NP_001028448.3 | 51 | 16321 | ||
| GenBank mRNA | BC058659.1 | - | 12 | 3080 | ||