siRNA Details
Gene Symbol: Ermard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034891.3 | NP_001030063.1 | 3, 4 | 488 | ||
NM_001039552.2 | NP_001034641.2 | 1421 | ||||
GenBank mRNA | AK019108.1 | - | 4, 5 | 576 | ||
AK160176.1 | - | 3, 4 | 489 | |||
BC150877.1 | - | 1482 | 9087788 |
