siRNA Details
Gene Symbol: Ermard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034891.3 | NP_001030063.1 | 6, 7 | 946 | ||
NM_001039552.2 | NP_001034641.2 | 1879 | ||||
GenBank mRNA | AK160176.1 | - | 6, 7 | 947 | ||
BC150877.1 | - | 1940 | 9087788 |
