siRNA Details
| Gene Symbol: Cdkl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033443.4 | NP_001028615.1 | 10 | 1156 | ||
| GenBank mRNA | AK157995.1 | - | 10 | 1157 | ||
| BC138986.1 | - | 9 | 1054 | 8862008 | ||
| BC138988.1 | - | 9 | 1054 | 8862010 | ||

Cdkl4
Genomic Map