siRNA Details
Gene Symbol: Arl14epl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033446.2 | NP_001028618.1 | 3 | 211 | ||
GenBank mRNA | AK145795.1 | - | 3 | 211 | ||
BC147441.1 | - | 2 | 55 | 9056083 | ||
BC147472.1 | - | 2 | 55 | 9056114 |
Gene Symbol: Gm7904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028798.1 | - | 1 | 19 |
