siRNA Details
| Gene Symbol: Arl14epl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033446.2 | NP_001028618.1 | 4 | 398 | ||
| GenBank mRNA | AK145795.1 | - | 4 | 398 | ||
| BC147441.1 | - | 3 | 242 | 9056083 | ||
| BC147472.1 | - | 3 | 242 | 9056114 | ||

Arl14epl
Genomic Map