siRNA Details
| Gene Symbol: 4430402I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198651.2 | NP_941053.2 | 10 | 1303 | ||
| GenBank mRNA | AK031997.1 | - | 10 | 1303 | ||
| BC147044.1 | - | 9 | 883 | 9056033 | ||
| BC147045.1 | - | 9 | 883 | 9056045 | ||
| BC171985.1 | - | 8 | 832 | 9054271 | ||
| BC171986.1 | - | 8 | 841 | 9054272 | ||

4430402I18Rik
Genomic Map