siRNA Details
| Gene Symbol: Fam178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126046.1 | NP_001119518.1 | 9 | 1288 | ||
| NM_201365.2 | NP_958753.2 | 9 | 1288 | |||
| GenBank mRNA | AK141223.1 | - | 9 | 1244 | ||
| BC050210.1 | - | 9 | 1191 | 30009484 | ||

Fam178b
Genomic Map