siRNA Details
| Gene Symbol: Trim55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081281.1 | NP_001074750.1 | 3 | 541 | ||
| GenBank mRNA | AK052918.1 | - | 3 | 542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim55| Gene Symbol: Trim55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081281.1 | NP_001074750.1 | 3 | 541 | ||
| GenBank mRNA | AK052918.1 | - | 3 | 542 | ||