siRNA Details
Gene Symbol: Tbc1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198664.3 | NP_941066.3 | 3 | 703 | ||
GenBank mRNA | AK041359.1 | - | 3 | 715 | ||
AK148254.1 | - | 3 | 706 | |||
AK148289.1 | - | 3 | 620 |
