siRNA Details
| Gene Symbol: Thap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039179.1 | - | 4 | 1133 | ||
| AK080650.1 | - | 4 | 1132 | |||
| BC089377.1 | - | 5 | 806 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thap6| Gene Symbol: Thap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039179.1 | - | 4 | 1133 | ||
| AK080650.1 | - | 4 | 1132 | |||
| BC089377.1 | - | 5 | 806 | |||