siRNA Details
| Gene Symbol: Gm1045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177577.1 | NP_001171048.1 | 2 | 212 | ||
| GenBank mRNA | CB233033.1 | - | 2 | 213 | ||
| KC602377.1 | - | 3 | 272 | |||
| KF477194.1 | - | 2 | 193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1045| Gene Symbol: Gm1045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177577.1 | NP_001171048.1 | 2 | 212 | ||
| GenBank mRNA | CB233033.1 | - | 2 | 213 | ||
| KC602377.1 | - | 3 | 272 | |||
| KF477194.1 | - | 2 | 193 | |||