siRNA Details
Gene Symbol: Lrcol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033459.3 | NP_001028631.1 | 2 | 245 | ||
GenBank mRNA | AK141249.1 | - | 2 | 246 | ||
BC147447.1 | - | 2 | 183 | 9056089 | ||
BC147469.1 | - | 2 | 183 | 9056111 | ||
BC171993.1 | - | 2 | 161 | |||
JQ258939.1 | - | 4 | 390 | |||
JQ258940.1 | - | 5 | 453 | |||
JQ258941.1 | - | 4 | 650 |
