siRNA Details
| Gene Symbol: Tatdn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033463.3 | NP_001028635.2 | 3 | 554 | ||
| GenBank mRNA | AK146408.1 | - | 2 | 303 | ||
| AK156197.1 | - | 3 | 555 | |||
| AK220217.1 | - | 2 | 485 | |||
| BC141065.1 | - | 3 | 489 | 9055677 | ||
| BC145318.1 | - | 3 | 489 | 9053873 | ||

Tatdn2
Genomic Map