siRNA Details
| Gene Symbol: Clec4b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004159.2 | NP_001004159.1 | 4 | 323 | ||
| GenBank mRNA | AK156711.1 | - | 4 | 455 | ||
| AY365133.1 | - | 4 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4b2| Gene Symbol: Clec4b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004159.2 | NP_001004159.1 | 4 | 323 | ||
| GenBank mRNA | AK156711.1 | - | 4 | 455 | ||
| AY365133.1 | - | 4 | 346 | |||