siRNA Details
| Gene Symbol: Lpar5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163268.1 | NP_001156740.1 | 2 | 417 | ||
| NM_001163269.1 | NP_001156741.1 | 2 | 352 | |||
| GenBank mRNA | AK131863.1 | - | 2 | 353 | ||
| AY255621.1 | - | 1 | 30 | |||
| BC116702.1 | - | 2 | 317 | 40104865 | ||
| BC117528.1 | - | 2 | 317 | 40104864 | ||
More Information
Set Membership: |
|

Lpar5
Genomic Map