siRNA Details
| Gene Symbol: 1190002F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI121041.1 | - | 2 | 153 | ||
| AK004447.1 | - | 2 | 193 | |||
| AK012387.1 | - | 2 | 131 | |||
| AK138072.1 | - | 3383 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1190002F15Rik| Gene Symbol: 1190002F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI121041.1 | - | 2 | 153 | ||
| AK004447.1 | - | 2 | 193 | |||
| AK012387.1 | - | 2 | 131 | |||
| AK138072.1 | - | 3383 | ||||