siRNA Details
| Gene Symbol: Ovol3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289817.1 | NP_001276746.1 | 3 | 1604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ovol3| Gene Symbol: Ovol3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289817.1 | NP_001276746.1 | 3 | 1604 | ||