siRNA Details
| Gene Symbol: 6430531B16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033465.2 | NP_001028637.1 | 4 | 631 | ||
| GenBank mRNA | AK078239.1 | - | 254 | |||
| AK131789.1 | - | 4 | 632 | |||
| AK164921.1 | - | 4 | 601 | |||
| BC025923.1 | - | 3 | 967 | |||
| BC145460.1 | - | 4 | 595 | |||
| BC145461.1 | - | 4 | 595 | |||
| BC145730.1 | - | 4 | 595 | 40131033 | ||
| BC171930.1 | - | 4 | 595 | |||

6430531B16Rik
Genomic Map