siRNA Details
| Gene Symbol: 6430531B16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033465.2 | NP_001028637.1 | 5, 6 | 769 | ||
| GenBank mRNA | AK131789.1 | - | 5, 6 | 770 | ||
| BC025923.1 | - | 3, 4 | 1013 | |||
| BC145460.1 | - | 4, 5 | 645 | |||
| BC145461.1 | - | 5, 6 | 729 | |||
| BC145730.1 | - | 5, 6 | 733 | 40131033 | ||

6430531B16Rik
Genomic Map