siRNA Details
| Gene Symbol: Mrgprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203492.2 | NP_987077.1 | 2 | 589 | ||
| GenBank mRNA | AJ313465.1 | - | 1 | 635 | ||
| BC100302.1 | - | 2 | 589 | 30522259 | ||
More Information
Set Membership: |
|

Mrgprg
Genomic Map