siRNA Details
| Gene Symbol: Mrgprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203492.2 | NP_987077.1 | 2 | 699 | ||
| GenBank mRNA | BC100302.1 | - | 2 | 699 | 30522259 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrgprg| Gene Symbol: Mrgprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203492.2 | NP_987077.1 | 2 | 699 | ||
| GenBank mRNA | BC100302.1 | - | 2 | 699 | 30522259 | |
Set Membership: |
|