siRNA Details
Gene Symbol: Urb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029876.1 | NP_001025047.1 | 1 | 57 | ||
GenBank mRNA | AK133906.1 | - | 2 | 371 | ||
BC151125.1 | - | 1 | 136 | 9088038 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Urb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029876.1 | NP_001025047.1 | 1 | 57 | ||
GenBank mRNA | AK133906.1 | - | 2 | 371 | ||
BC151125.1 | - | 1 | 136 | 9088038 |