siRNA Details
| Gene Symbol: Ccdc84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201372.3 | NP_958760.2 | 10 | 904 | ||
| GenBank mRNA | BC055929.1 | - | 10 | 904 | ||
| BC096560.1 | - | 10 | 909 | 1527981 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc84| Gene Symbol: Ccdc84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201372.3 | NP_958760.2 | 10 | 904 | ||
| GenBank mRNA | BC055929.1 | - | 10 | 904 | ||
| BC096560.1 | - | 10 | 909 | 1527981 | ||