siRNA Details
| Gene Symbol: Fbxw26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198674.2 | NP_941076.1 | 3 | 208 | ||
| GenBank mRNA | AK054298.1 | - | 3 | 208 | ||
| BC132114.1 | - | 3 | 207 | 40130391 | ||
| BC132304.1 | - | 3 | 207 | 40130581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw26| Gene Symbol: Fbxw26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198674.2 | NP_941076.1 | 3 | 208 | ||
| GenBank mRNA | AK054298.1 | - | 3 | 208 | ||
| BC132114.1 | - | 3 | 207 | 40130391 | ||
| BC132304.1 | - | 3 | 207 | 40130581 | ||