siRNA Details
| Gene Symbol: Fbxw22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014395.2 | NP_001014417.1 | 6 | 795 | ||
| GenBank mRNA | AK135808.1 | - | 4 | 512 | ||
| AK162068.1 | - | 2 | 303 | |||
| BC084593.1 | - | 6 | 799 | 30477093 | ||
| CJ052642.1 | - | 4 | 512 | |||

Fbxw22
Genomic Map