siRNA Details
| Gene Symbol: Gm1140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126317.1 | NP_001119789.1 | 7, 8 | 625 | ||
| Gene Symbol: Gm14692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163195.1 | NP_001156667.1 | 7, 8 | 625 | ||

Gm1140
Genomic Map