siRNA Details
| Gene Symbol: Tmem29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164686.1 | NP_001158158.1 | 3 | 460 | ||
| GenBank mRNA | AK012546.1 | - | 3 | 471 | ||
| BC058428.1 | - | 3 | 421 | |||
| CA526769.1 | - | 3 | 461 | |||
| CK624690.1 | - | 3 | 431 | |||

Tmem29
Genomic Map