siRNA Details
| Gene Symbol: Atxn7l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033474.2 | NP_001028646.2 | 1 | 367 | ||
| GenBank mRNA | AK003631.1 | - | 1 | 337 | ||
| AK009409.1 | - | 1 | 348 | |||
| AK011696.1 | - | 1 | 341 | |||
| AK014824.1 | - | 1 | 263 | |||
| AK135540.1 | - | 1 | 340 | |||
| AK150316.1 | - | 1 | 337 | |||
| BC030168.1 | - | 1 | 336 | |||
| BC047210.1 | - | 1 | 337 | |||
| BC058561.1 | - | 1 | 299 | |||
| BC079579.1 | - | 1 | 322 | |||
| BC094390.1 | - | 1 | 325 | |||
| Gene Symbol: Gm26596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143218.1 | - | 1 | 157 | ||

Atxn7l3b
Genomic Map