siRNA Details
Gene Symbol: Ypel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027166.5 | NP_081442.1 | 3 | 346 | ||
GenBank mRNA | AB098744.1 | - | 2 | 234 | ||
AK010201.1 | - | 3 | 312 | |||
AK046281.1 | - | 3 | 345 | |||
AK049096.1 | - | 3 | 347 | |||
AK049644.1 | - | 3 | 345 | |||
AK137446.1 | - | 3 | 318 | |||
AK154461.1 | - | 3 | 385 | |||
AK165943.1 | - | 3 | 348 | |||
AK167998.1 | - | 3 | 362 | |||
AK169241.1 | - | 3 | 349 | |||
AK169986.1 | - | 3 | 320 | |||
AK171010.1 | - | 3 | 347 | |||
BC025125.1 | - | 1 | 106 | 4921751 | ||
BC085109.1 | - | 3 | 365 | 30737658 |
