siRNA Details
| Gene Symbol: Fam47e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033478.2 | NP_001028650.2 | 5 | 583 | ||
| NM_001170571.1 | NP_001164042.1 | 3 | 400 | |||
| GenBank mRNA | AA244788.1 | - | 3 | 381 | ||
| AK144479.1 | - | 5 | 583 | |||
| BC147167.1 | - | 1 | 81 | 9056206 | ||
| BC147168.1 | - | 1 | 81 | 9056207 | ||
| BE650192.1 | - | 5 | 487 | |||

Fam47e
Genomic Map