siRNA Details
| Gene Symbol: Fam47e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033478.2 | NP_001028650.2 | 6 | 699 | ||
| NM_001170571.1 | NP_001164042.1 | 4 | 516 | |||
| GenBank mRNA | AK050399.1 | - | 2 | 217 | ||
| AK144479.1 | - | 6 | 699 | |||
| BC147167.1 | - | 2 | 197 | 9056206 | ||
| BC147168.1 | - | 2 | 197 | 9056207 | ||

Fam47e
Genomic Map