siRNA Details
| Gene Symbol: Zfp667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024928.2 | NP_001020099.1 | 5 | 599 | ||
| GenBank mRNA | AK080616.1 | - | 5 | 578 | ||
| AK164056.1 | - | 6 | 508 | |||
| AY850275.1 | - | 5 | 599 | |||
| BC049673.1 | - | 5 | 599 | |||
| BC141152.1 | - | 5 | 634 | 9055764 | ||
| BC141153.1 | - | 5 | 634 | 9055765 | ||

Zfp667
Genomic Map