siRNA Details
| Gene Symbol: Gm5346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025240.1 | NP_001020411.1 | 1 | 2167 | ||
| GenBank mRNA | BC151154.1 | - | 1 | 2259 | 9087881 | |
| BC157897.1 | - | 1 | 2259 | 9087877 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5346| Gene Symbol: Gm5346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025240.1 | NP_001020411.1 | 1 | 2167 | ||
| GenBank mRNA | BC151154.1 | - | 1 | 2259 | 9087881 | |
| BC157897.1 | - | 1 | 2259 | 9087877 | ||