siRNA Details
| Gene Symbol: Gm1527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033479.4 | NP_001028651.1 | 9 | 1025 | ||
| GenBank mRNA | AK132999.1 | - | 9 | 1024 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1527| Gene Symbol: Gm1527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033479.4 | NP_001028651.1 | 9 | 1025 | ||
| GenBank mRNA | AK132999.1 | - | 9 | 1024 | ||