siRNA Details
| Gene Symbol: 4930557A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029229.1 | NP_083505.1 | 1 | 163 | ||
| GenBank mRNA | AK016155.1 | - | 1 | 164 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930557A04Rik| Gene Symbol: 4930557A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029229.1 | NP_083505.1 | 1 | 163 | ||
| GenBank mRNA | AK016155.1 | - | 1 | 164 | ||