siRNA Details
| Gene Symbol: Frmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190332.1 | NP_001177261.1 | 12 | 2002 | ||
| GenBank mRNA | AK032347.1 | - | 1 | 333 | ||
| BC147257.1 | - | 1 | 277 | 9056311 | ||
| BC147258.1 | - | 1 | 277 | 9056312 | ||

Frmd7
Genomic Map