siRNA Details
| Gene Symbol: Kng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102409.1 | NP_001095879.1 | 4 | 718 | ||
| NM_001102410.1 | NP_001095880.1 | 4 | 718 | |||
| NM_201375.2 | NP_958763.1 | 4 | 718 | |||
| GenBank mRNA | AY462056.1 | - | 4 | 634 | ||
| AY462057.1 | - | 4 | 634 | |||
| AY462059.1 | - | 4 | 634 | |||
| AY660572.1 | - | 4 | 634 | |||
| BC108335.1 | - | 4 | 685 | 5124483 | ||
| BC139720.1 | - | 4 | 626 | 40039854 | ||

Kng2
Genomic Map