siRNA Details
| Gene Symbol: Slitrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198864.2 | NP_942564.2 | 2 | 1559 | ||
| GenBank mRNA | AK139378.1 | - | 2 | 1682 | ||
| AK139438.1 | - | 2 | 1336 | |||
| AK158856.1 | - | 2 | 1414 | |||
| BC066059.1 | - | 2 | 1559 | 6844461 | ||

Slitrk3
Genomic Map