siRNA Details
| Gene Symbol: Tas2r135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199159.1 | NP_954610.1 | 1 | 492 | ||
| GenBank mRNA | BC115532.1 | - | 1 | 488 | 40101425 | |
| BC115533.1 | - | 1 | 488 | 40101426 | ||
| BC139470.1 | - | 1 | 587 | 8862492 | ||
| BC139473.1 | - | 1 | 587 | 8862495 | ||

Tas2r135
Genomic Map