siRNA Details
| Gene Symbol: Znrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199143.2 | NP_954594.1 | 2 | 606 | ||
| GenBank mRNA | AF513708.1 | - | 2 | 606 | ||
| BC104316.2 | - | 2 | 251 | |||
| BC104317.2 | - | 2 | 260 | |||
| BC125242.1 | - | 2 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znrf2| Gene Symbol: Znrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199143.2 | NP_954594.1 | 2 | 606 | ||
| GenBank mRNA | AF513708.1 | - | 2 | 606 | ||
| BC104316.2 | - | 2 | 251 | |||
| BC104317.2 | - | 2 | 260 | |||
| BC125242.1 | - | 2 | 260 | |||