siRNA Details
| Gene Symbol: Banf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044750.1 | NP_001038215.1 | 2 | 174 | ||
| NM_207275.1 | NP_997158.1 | 3 | 294 | |||
| GenBank mRNA | AK076863.1 | - | 3 | 294 | ||
| BC049628.1 | - | 174 | 6706626 | |||

Banf2
Genomic Map