siRNA Details
Gene Symbol: Ccdc121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207280.3 | NP_997163.2 | 1 | 985 | ||
GenBank mRNA | AK078370.1 | - | 1 | 456 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207280.3 | NP_997163.2 | 1 | 985 | ||
GenBank mRNA | AK078370.1 | - | 1 | 456 |