siRNA Details
| Gene Symbol: 4930504O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207527.3 | NP_997410.1 | 3 | 176 | ||
| GenBank mRNA | AK076838.1 | - | 3 | 134 | ||
| BC048595.1 | - | 3 | 137 | 6740346 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930504O13Rik| Gene Symbol: 4930504O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207527.3 | NP_997410.1 | 3 | 176 | ||
| GenBank mRNA | AK076838.1 | - | 3 | 134 | ||
| BC048595.1 | - | 3 | 137 | 6740346 | ||