siRNA Details
| Gene Symbol: Olfr592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207556.2 | NP_997439.2 | 1 | 31 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr592| Gene Symbol: Olfr592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207556.2 | NP_997439.2 | 1 | 31 | ||