siRNA Details
| Gene Symbol: Olfr750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207558.2 | NP_997441.2 | 1 | 865 | ||
| GenBank mRNA | BC145881.1 | - | 1 | 865 | 40131184 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr750| Gene Symbol: Olfr750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207558.2 | NP_997441.2 | 1 | 865 | ||
| GenBank mRNA | BC145881.1 | - | 1 | 865 | 40131184 | |