siRNA Details
Gene Symbol: Olfr1383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207574.1 | NP_997457.1 | 1 | 903 | ||
GenBank mRNA | BC127965.1 | - | 1 | 907 | 40134985 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207574.1 | NP_997457.1 | 1 | 903 | ||
GenBank mRNA | BC127965.1 | - | 1 | 907 | 40134985 |