siRNA Details
| Gene Symbol: BC040756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040756.1 | - | 169 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC040756| Gene Symbol: BC040756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040756.1 | - | 169 | |||