siRNA Details
| Gene Symbol: Zyg11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033634.3 | NP_001028806.2 | 11 | 1985 | ||
| GenBank mRNA | AK122545.1 | - | 4 | 374 | ||
| AK145130.1 | - | 11 | 1959 | |||
| AK148335.1 | - | 11 | 1984 | |||
| AK148760.1 | - | 5 | 455 | |||
| AK166031.1 | - | 11 | 1965 | |||
| AK166174.1 | - | 11 | 1965 | |||
| AK166278.1 | - | 11 | 1976 | |||
| BC139383.1 | - | 11 | 1952 | 8862405 | ||
| BC139384.1 | - | 11 | 1952 | 8862406 | ||

Zyg11b
Genomic Map