siRNA Details
| Gene Symbol: Hnrnph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079824.1 | NP_001073293.1 | 2 | 227 | ||
| GenBank mRNA | BC049652.1 | - | 2 | 225 | ||
| BC147496.1 | - | 2 | 208 | |||
| BQ443117.1 | - | 1 | 68 | |||
| BU946493.1 | - | 2 | 228 | |||
| Gene Symbol: Aff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC094538.1 | - | 68 | |||

Hnrnph3
Genomic Map